Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAGGACAGTTTGACTTTGCACA[C/G]AAAGGAAAATCATATCTAATTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSMD1 - CUB and Sushi multiple domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033225.5 | 10590 | UTR 3 | NP_150094.5 | |||
XM_011534752.2 | 10590 | UTR 3 | XP_011533054.1 | |||
XM_011534753.2 | 10590 | UTR 3 | XP_011533055.1 | |||
XM_011534754.1 | 10590 | UTR 3 | XP_011533056.1 | |||
XM_017013731.1 | 10590 | Intron | XP_016869220.1 |