Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTAACTGATGGCCACGGTCCC[C/T]GTTCAGAACCTTCTGTGTCCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616748 MIM: 611255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTPD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENTPD8 - ectonucleoside triphosphate diphosphohydrolase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EXD3 - exonuclease 3'-5' domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOXA1 - NADPH oxidase activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256067.1 | Intron | NP_001242996.1 | ||||
NM_001256068.1 | Intron | NP_001242997.1 | ||||
NM_006647.1 | Intron | NP_006638.1 | ||||
XM_011518158.2 | Intron | XP_011516460.1 | ||||
XM_011518159.1 | Intron | XP_011516461.1 | ||||
XM_011518160.1 | Intron | XP_011516462.1 | ||||
XM_017014220.1 | Intron | XP_016869709.1 |