Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTGGCTCCTGGGCATCTCTGT[G/T]CCTCTGTCTCCCACATGTTAGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1B - calcium voltage-gated channel subunit alpha1 B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000718.3 | Intron | NP_000709.1 | ||||
NM_001243812.1 | Intron | NP_001230741.1 | ||||
XM_011518991.2 | Intron | XP_011517293.1 | ||||
XM_011518992.2 | Intron | XP_011517294.1 | ||||
XM_011518993.2 | Intron | XP_011517295.1 | ||||
XM_011518994.2 | Intron | XP_011517296.1 | ||||
XM_017015115.1 | Intron | XP_016870604.1 | ||||
XM_017015116.1 | Intron | XP_016870605.1 | ||||
XM_017015117.1 | Intron | XP_016870606.1 | ||||
XM_017015118.1 | Intron | XP_016870607.1 | ||||
XM_017015119.1 | Intron | XP_016870608.1 | ||||
XM_017015120.1 | Intron | XP_016870609.1 |
LOC100133077 - uncharacterized LOC100133077 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |