Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCGTGAAGCTAATCCTCGAGA[C/T]TCCAGTTGAATATTTCTCTCAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603651 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPC4 - transient receptor potential cation channel subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135955.1 | 3457 | Silent Mutation | GAA,GAG | E,E 803 | NP_001129427.1 | |
NM_001135956.1 | 3457 | Silent Mutation | GAA,GAG | E,E 738 | NP_001129428.1 | |
NM_001135957.1 | 3457 | Silent Mutation | GAA,GAG | E,E 746 | NP_001129429.1 | |
NM_001135958.1 | 3457 | Silent Mutation | GAA,GAG | E,E 714 | NP_001129430.1 | |
NM_003306.1 | 3457 | Silent Mutation | GAA,GAG | E,E 892 | NP_003297.1 | |
NM_016179.2 | 3457 | Silent Mutation | GAA,GAG | E,E 887 | NP_057263.1 | |
XM_011535206.1 | 3457 | Silent Mutation | GAA,GAG | E,E 472 | XP_011533508.1 | |
XM_017020723.1 | 3457 | Silent Mutation | GAA,GAG | E,E 472 | XP_016876212.1 | |
XM_017020724.1 | 3457 | Silent Mutation | GAA,GAG | E,E 467 | XP_016876213.1 |