Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCAGTAGAGATTAAATATGAG[G/T]TCAGGAATCAGCTCCTCCAAACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607441 | ||||||||||||||||||||
Literature Links: |
EPSTI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPSTI1 - epithelial stromal interaction 1 (breast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266596.1 | 1667 | Silent Mutation | XP_005266653.1 | |||
XM_005266597.1 | 1667 | UTR 3 | XP_005266654.1 | |||
XM_006719896.3 | 1667 | Intron | XP_006719959.1 | |||
XM_011535312.1 | 1667 | Silent Mutation | XP_011533614.1 | |||
XM_011535313.1 | 1667 | Silent Mutation | XP_011533615.1 | |||
XM_011535314.1 | 1667 | Silent Mutation | XP_011533616.1 | |||
XM_017020853.1 | 1667 | Silent Mutation | XP_016876342.1 | |||
XM_017020854.1 | 1667 | Silent Mutation | XP_016876343.1 | |||
XM_017020855.1 | 1667 | Silent Mutation | XP_016876344.1 |