Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGGCATGATGACTCTGCAGGC[C/T]GAACGGCAGCCTTGATAATTATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605147 MIM: 604098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LECT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LECT1 - leukocyte cell derived chemotaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011705.1 | 940 | Silent Mutation | TCA,TCG | S,S 314 | NP_001011705.1 | |
NM_007015.2 | 940 | Silent Mutation | TCA,TCG | S,S 315 | NP_008946.1 | |
XM_011534897.2 | 940 | Silent Mutation | TCA,TCG | S,S 346 | XP_011533199.1 | |
XM_011534898.2 | 940 | Silent Mutation | TCA,TCG | S,S 345 | XP_011533200.1 | |
XM_011534899.2 | 940 | Silent Mutation | TCA,TCG | S,S 308 | XP_011533201.1 | |
XM_011534900.2 | 940 | Silent Mutation | TCA,TCG | S,S 304 | XP_011533202.1 |
SUGT1 - SGT1 homolog, MIS12 kinetochore complex assembly cochaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |