Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGATGTTTTTAAAGTTAAAAAA[A/T]CTCCACTACATGCCATATTGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604490 MIM: 608896 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SACS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SACS - sacsin molecular chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278055.1 | 14740 | UTR 3 | NP_001264984.1 | |||
NM_014363.5 | 14740 | UTR 3 | NP_055178.3 | |||
XM_005266338.2 | 14740 | UTR 3 | XP_005266395.1 | |||
XM_011535039.2 | 14740 | UTR 3 | XP_011533341.1 | |||
XM_017020539.1 | 14740 | UTR 3 | XP_016876028.1 |
SGCG - sarcoglycan gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |