Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGGCACCACCTCCTCCTCTAG[G/T]GACTCCACGTAGAAGAGAGTCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603680 MIM: 605332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN8OS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATXN8OS - ATXN8 opposite strand (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHL1 - kelch like family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286725.1 | 1166 | Silent Mutation | TCA,TCC | S,S 124 | NP_001273654.1 | |
NM_020866.2 | 1166 | Silent Mutation | TCA,TCC | S,S 124 | NP_065917.1 | |
XM_017020678.1 | 1166 | Intron | XP_016876167.1 | |||
XM_017020679.1 | 1166 | Intron | XP_016876168.1 |