Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATGTGAAAAGGAAACACACAC[A/G]CAGATGTTTGCCTGGAAATGCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLT1 - fms related tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159920.1 | Intron | NP_001153392.1 | ||||
NM_001160030.1 | Intron | NP_001153502.1 | ||||
NM_001160031.1 | Intron | NP_001153503.1 | ||||
NM_002019.4 | Intron | NP_002010.2 | ||||
XM_011535014.1 | Intron | XP_011533316.1 | ||||
XM_017020485.1 | Intron | XP_016875974.1 |
PAN3 - PAN3 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |