Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTTAAGGCCAGAGAGTGGCTA[A/C]TTCTCTGCCTCCTGCAGCTCCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614544 | ||||||||||||||||||||
Literature Links: |
FAM69C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM69C - family with sequence similarity 69 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044369.2 | 1312 | Missense Mutation | AAG,AAT | K,N 419 | NP_001037834.2 | |
XM_011525816.2 | 1312 | Missense Mutation | AAG,AAT | K,N 381 | XP_011524118.1 | |
XM_017025551.1 | 1312 | Missense Mutation | AAG,AAT | K,N 278 | XP_016881040.1 |