Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTAATTTTATTGTGATGTTATG[C/T]CTCAAAATGTAATACAAAATTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603049 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNL1 - thioredoxin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004786.2 | 1328 | Intron | NP_004777.1 | |||
XM_006722580.3 | 1328 | Intron | XP_006722643.1 | |||
XM_011526263.2 | 1328 | UTR 3 | XP_011524565.1 | |||
XM_011526264.2 | 1328 | Intron | XP_011524566.1 | |||
XM_017026093.1 | 1328 | Intron | XP_016881582.1 | |||
XM_017026094.1 | 1328 | UTR 3 | XP_016881583.1 |