Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTAATGGCAAAGGCTAGAGGAA[A/G]ATCCTAAACAATCACCTATGCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FECH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FECH - ferrochelatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000140.3 | 3029 | UTR 3 | NP_000131.2 | |||
NM_001012515.2 | 3029 | UTR 3 | NP_001012533.1 | |||
XM_011525881.1 | 3029 | UTR 3 | XP_011524183.1 | |||
XM_011525882.2 | 3029 | UTR 3 | XP_011524184.1 | |||
XM_017025614.1 | 3029 | Intron | XP_016881103.1 |