Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAAGAGGGCCAGAGAACCCACC[A/G]CTAAAGCCGTAGGGTGGGAGGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF152 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF152 - ring finger protein 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173557.2 | 8534 | Intron | NP_775828.1 | |||
XM_005266650.3 | 8534 | UTR 3 | XP_005266707.1 | |||
XM_005266652.3 | 8534 | UTR 3 | XP_005266709.1 | |||
XM_011525878.2 | 8534 | UTR 3 | XP_011524180.1 | |||
XM_011525879.2 | 8534 | UTR 3 | XP_011524181.1 | |||
XM_017025612.1 | 8534 | UTR 3 | XP_016881101.1 | |||
XM_017025613.1 | 8534 | UTR 3 | XP_016881102.1 |