Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCAGCAGCCACAGGAGAGACAG[A/G]AAGAAAACCAAGAGTAAGGTGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf203 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf203 - chromosome 6 open reading frame 203 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142468.2 | Intron | NP_001135940.1 | ||||
NM_001142470.2 | Intron | NP_001135942.1 | ||||
NM_016487.4 | Intron | NP_057571.1 | ||||
XM_011535876.2 | Intron | XP_011534178.1 | ||||
XM_011535877.2 | Intron | XP_011534179.1 | ||||
XM_017010919.1 | Intron | XP_016866408.1 | ||||
XM_017010920.1 | Intron | XP_016866409.1 |