Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTAGGTAATCAGTAGCTGCCTCA[A/G]GACACTGGTGCTTCACAGCAGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC63 - SEC63 homolog, protein translocation regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007214.4 | 4164 | UTR 3 | NP_009145.1 | |||
XM_011535399.2 | 4164 | Intron | XP_011533701.1 | |||
XM_017010218.1 | 4164 | Intron | XP_016865707.1 |