Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGTGGTTGCTGGTATTGGTCCC[A/G]CAGCTCAAGAGACTGGGGCTGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD164 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD164 - CD164 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL6 - peptidylprolyl isomerase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111298.2 | 2112 | UTR 3 | NP_001104768.2 | |||
NM_001286360.1 | 2112 | UTR 3 | NP_001273289.1 | |||
NM_001286361.1 | 2112 | UTR 3 | NP_001273290.1 | |||
NM_173672.4 | 2112 | UTR 3 | NP_775943.1 | |||
XM_011535765.2 | 2112 | Intron | XP_011534067.1 | |||
XM_011535766.2 | 2112 | Intron | XP_011534068.1 | |||
XM_011535767.2 | 2112 | Intron | XP_011534069.1 | |||
XM_011535769.2 | 2112 | Intron | XP_011534071.1 | |||
XM_017010774.1 | 2112 | Intron | XP_016866263.1 |