Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAAATAATTCAGGCATAGCTAG[A/G]ACTGATGCTCAAGTTAGAAACAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607129 MIM: 614064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICAL1 - microtubule associated monooxygenase, calponin and LIM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159291.1 | 4703 | Intron | NP_001152763.1 | |||
NM_001286613.1 | 4703 | Intron | NP_001273542.1 | |||
NM_022765.3 | 4703 | Intron | NP_073602.3 |
ZBTB24 - zinc finger and BTB domain containing 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164313.1 | 4703 | Intron | NP_001157785.1 | |||
NM_014797.2 | 4703 | UTR 3 | NP_055612.2 |