Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTAATGGCTTTGCTTTGACAAC[A/G]CTTTTCCCATAGGGGAAATGGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMD1 - adenosylmethionine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287214.1 | Intron | NP_001274143.1 | ||||
NM_001287215.1 | Intron | NP_001274144.1 | ||||
NM_001287216.1 | Intron | NP_001274145.1 | ||||
NM_001634.5 | Intron | NP_001625.2 |