Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGTATTAAGGTCTTCATATCCC[A/G]TAACCCGCTTTGCATCACCTGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606914 MIM: 614548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKIB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKIB - protein kinase (cAMP-dependent, catalytic) inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270393.1 | Intron | NP_001257322.1 | ||||
NM_001270394.1 | Intron | NP_001257323.1 | ||||
NM_001270395.1 | Intron | NP_001257324.1 | ||||
NM_032471.5 | Intron | NP_115860.1 | ||||
NM_181794.2 | Intron | NP_861459.1 | ||||
NM_181795.2 | Intron | NP_861460.1 | ||||
XM_011535930.2 | Intron | XP_011534232.1 | ||||
XM_011535931.2 | Intron | XP_011534233.1 | ||||
XM_011535932.2 | Intron | XP_011534234.1 | ||||
XM_011535933.2 | Intron | XP_011534235.1 | ||||
XM_011535934.2 | Intron | XP_011534236.1 | ||||
XM_011535935.2 | Intron | XP_011534237.1 | ||||
XM_011535936.1 | Intron | XP_011534238.1 | ||||
XM_011535937.2 | Intron | XP_011534239.1 |
SERINC1 - serine incorporator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |