Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGGGCTAAAAGAAATGAAATTA[C/T]ATAGAAGTAAAATGTAAAAGACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ3 - coenzyme Q3, methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017421.3 | Intron | NP_059117.3 | ||||
XM_005267018.4 | Intron | XP_005267075.1 | ||||
XM_006715500.3 | Intron | XP_006715563.1 | ||||
XM_011535898.2 | Intron | XP_011534200.1 | ||||
XM_011535899.1 | Intron | XP_011534201.1 | ||||
XM_017010933.1 | Intron | XP_016866422.1 |