Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGTTTTGTGTGAAATAAACATTT[G/T]GCTGAAGTGCAATAGCTGCTGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602448 MIM: 604108 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K5 - mitogen-activated protein kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005923.3 | 4373 | UTR 3 | NP_005914.1 | |||
XM_011535839.2 | 4373 | Intron | XP_011534141.2 | |||
XM_017010870.1 | 4373 | Intron | XP_016866359.1 | |||
XM_017010871.1 | 4373 | Intron | XP_016866360.1 | |||
XM_017010872.1 | 4373 | UTR 3 | XP_016866361.1 | |||
XM_017010873.1 | 4373 | UTR 3 | XP_016866362.1 | |||
XM_017010874.1 | 4373 | Intron | XP_016866363.1 | |||
XM_017010875.1 | 4373 | UTR 3 | XP_016866364.1 | |||
XM_017010876.1 | 4373 | Intron | XP_016866365.1 | |||
XM_017010877.1 | 4373 | UTR 3 | XP_016866366.1 |
MAP7 - microtubule associated protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |