Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCAACAGCATCTTTTTGACAA[A/G]AAGACCTAGCCACAGACAAGGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614725 | ||||||||||||||||||||
Literature Links: |
SERAC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERAC1 - serine active site containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032861.3 | 1574 | Missense Mutation | CTT,TTT | L,F 503 | NP_116250.3 | |
XM_006715586.2 | 1574 | Missense Mutation | CTT,TTT | L,F 433 | XP_006715649.1 | |
XM_011536196.2 | 1574 | Missense Mutation | CTT,TTT | L,F 496 | XP_011534498.1 | |
XM_011536198.2 | 1574 | Missense Mutation | CTT,TTT | L,F 433 | XP_011534500.1 |