Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTAGATACCTGAGATGACAAAA[C/T]TTTTATCAAACTTCATGTATTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR1OP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFR1OP - FGFR1 oncogene partner | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278690.1 | Intron | NP_001265619.1 | ||||
NM_007045.3 | Intron | NP_008976.1 | ||||
NM_194429.2 | Intron | NP_919410.1 |
MIR3939 - microRNA 3939 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |