Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCACAGCTAGTCCCCCACTCTCC[A/G]TCTGTCACCAAAGCTTCGCCCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD1 - FERM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122841.2 | 2380 | UTR 3 | NP_001116313.1 | |||
NM_024919.4 | 2380 | UTR 3 | NP_079195.3 | |||
XM_011536137.1 | 2380 | UTR 3 | XP_011534439.1 | |||
XM_011536138.1 | 2380 | UTR 3 | XP_011534440.1 | |||
XM_011536140.1 | 2380 | UTR 3 | XP_011534442.1 | |||
XM_011536141.1 | 2380 | UTR 3 | XP_011534443.1 | |||
XM_011536142.2 | 2380 | UTR 3 | XP_011534444.1 | |||
XM_011536143.1 | 2380 | UTR 3 | XP_011534445.1 | |||
XM_011536145.1 | 2380 | UTR 3 | XP_011534447.1 | |||
XM_011536146.2 | 2380 | UTR 3 | XP_011534448.1 | |||
XM_017011317.1 | 2380 | Intron | XP_016866806.1 | |||
XM_017011318.1 | 2380 | Intron | XP_016866807.1 |