Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTGCAAAACTAGAGCTATAAA[A/G]TCTAAGTTAAAAAATCAGCTTATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616987 MIM: 613069 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf120 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C6orf120 - chromosome 6 open reading frame 120 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029863.2 | 4066 | UTR 3 | NP_001025034.1 | |||
NM_001317342.1 | 4066 | UTR 3 | NP_001304271.1 |
PHF10 - PHD finger protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018288.3 | 4066 | Intron | NP_060758.2 | |||
NM_133325.2 | 4066 | Intron | NP_579866.2 | |||
XM_017010998.1 | 4066 | Intron | XP_016866487.1 |
WDR27 - WD repeat domain 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |