Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGGCCACAGAAGACATTTCCT[C/T]CTTCTTTAACCCTCAGAAAGAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607725 MIM: 601686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP2 - poly(ADP-ribose) polymerase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEP1 - telomerase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319035.1 | 8021 | UTR 3 | NP_001305964.1 | |||
NM_007110.4 | 8021 | UTR 3 | NP_009041.2 | |||
XM_005268027.4 | 8021 | UTR 3 | XP_005268084.1 | |||
XM_011537110.2 | 8021 | UTR 3 | XP_011535412.1 |