Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTTGCATTTCAGTATGTTCTA[C/G]AGCTTCTGTATCATCACCTGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAPPC6B - trafficking protein particle complex 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079537.1 | 809 | Silent Mutation | CTC,CTG | L,L 158 | NP_001073005.1 | |
NM_177452.3 | 809 | Silent Mutation | CTC,CTG | L,L 130 | NP_803235.1 | |
XM_011536409.2 | 809 | Silent Mutation | CTC,CTG | L,L 120 | XP_011534711.1 | |
XM_017020964.1 | 809 | Intron | XP_016876453.1 |