Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGTTATGCTCAACTAAACCTT[G/T]AGTTTTAAGATGTTTTATATTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186947 MIM: 614907 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP3 - FK506 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002013.3 | Intron | NP_002004.1 | ||||
XM_011536565.2 | Intron | XP_011534867.1 | ||||
XM_017021092.1 | Intron | XP_016876581.1 | ||||
XM_017021093.1 | Intron | XP_016876582.1 |
PRPF39 - pre-mRNA processing factor 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD127 - small nucleolar RNA, C/D box 127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |