Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGGCTTGGACAATTCTGGACA[A/G]TGACACAGGAGGAAAAGTCTGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606439 MIM: 604923 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATL1 - atlastin GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127713.1 | Intron | NP_001121185.1 | ||||
NM_015915.4 | Intron | NP_056999.2 | ||||
NM_181598.3 | Intron | NP_853629.2 |
MAP4K5 - mitogen-activated protein kinase kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006575.4 | Intron | NP_006566.2 | ||||
NM_198794.2 | Intron | NP_942089.1 | ||||
XM_006720013.3 | Intron | XP_006720076.1 | ||||
XM_011536377.2 | Intron | XP_011534679.1 | ||||
XM_011536378.2 | Intron | XP_011534680.1 | ||||
XM_011536379.2 | Intron | XP_011534681.1 | ||||
XM_011536380.2 | Intron | XP_011534682.1 |