Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCCTCAGTATAGTATTGTCTT[C/T]CTTATTCTTCTTTCTTTTTTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD12B - abhydrolase domain containing 12B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYGL - phosphorylase, glycogen, liver | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163940.1 | Intron | NP_001157412.1 | ||||
NM_002863.4 | Intron | NP_002854.3 |