Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTATGTTTGGCAAGGTCAAGT[A/G]AGATGGCTGAGAAGTATTCACTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR135 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR135 - G protein-coupled receptor 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022571.5 | 6587 | Intron | NP_072093.2 | |||
XM_017021596.1 | 6587 | UTR 3 | XP_016877085.1 | |||
XM_017021597.1 | 6587 | UTR 3 | XP_016877086.1 | |||
XM_017021598.1 | 6587 | UTR 3 | XP_016877087.1 | |||
XM_017021599.1 | 6587 | UTR 3 | XP_016877088.1 |