Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCTGGAACCACGGGCCTACAA[A/G]GGGAGGGGTTTAATCTAAAGTACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHG3 - pleckstrin homology and RhoGEF domain containing G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308147.1 | Intron | NP_001295076.1 | ||||
XM_011536624.2 | Intron | XP_011534926.1 | ||||
XM_011536625.2 | Intron | XP_011534927.1 | ||||
XM_011536627.2 | Intron | XP_011534929.1 | ||||
XM_011536628.2 | Intron | XP_011534930.1 | ||||
XM_011536629.2 | Intron | XP_011534931.1 | ||||
XM_017021154.1 | Intron | XP_016876643.1 | ||||
XM_017021155.1 | Intron | XP_016876644.1 | ||||
XM_017021156.1 | Intron | XP_016876645.1 | ||||
XM_017021157.1 | Intron | XP_016876646.1 | ||||
XM_017021158.1 | Intron | XP_016876647.1 |