Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCTTCCTTTCTCGGGTCTGTG[A/G]AGTTGCTTGCTGGAAATGCCTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHG3 - pleckstrin homology and RhoGEF domain containing G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308147.1 | 9413 | Intron | NP_001295076.1 |
SPTB - spectrin beta, erythrocytic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000347.5 | 9413 | Intron | NP_000338.3 | |||
NM_001024858.2 | 9413 | UTR 3 | NP_001020029.1 | |||
XM_005268023.4 | 9413 | UTR 3 | XP_005268080.1 | |||
XM_011537105.2 | 9413 | Intron | XP_011535407.1 | |||
XM_017021612.1 | 9413 | UTR 3 | XP_016877101.1 | |||
XM_017021613.1 | 9413 | Intron | XP_016877102.1 | |||
XM_017021614.1 | 9413 | Intron | XP_016877103.1 |