Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGTGTCCTGTAGGATAATGA[C/T]GGCCCCTCATGACTTGTCCTGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXD2 - exonuclease 3'-5' domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193360.1 | Intron | NP_001180289.1 | ||||
NM_001193361.1 | Intron | NP_001180290.1 | ||||
NM_001193362.1 | Intron | NP_001180291.1 | ||||
NM_001193363.1 | Intron | NP_001180292.1 | ||||
NM_018199.3 | Intron | NP_060669.1 | ||||
XM_005267817.3 | Intron | XP_005267874.1 | ||||
XM_011536908.2 | Intron | XP_011535210.1 | ||||
XM_017021421.1 | Intron | XP_016876910.1 |
LOC105370550 - uncharacterized LOC105370550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |