Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGACACAACAAGACACACAGAAT[C/T]GGAAAGGCTTGGGCAAAGAGGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC8A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC8A3 - solute carrier family 8 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130417.2 | 2083 | UTR 3 | NP_001123889.1 | |||
NM_033262.4 | 2083 | UTR 3 | NP_150287.1 | |||
NM_058240.3 | 2083 | UTR 3 | NP_489479.1 | |||
NM_182932.2 | 2083 | UTR 3 | NP_891977.1 | |||
NM_182936.2 | 2083 | UTR 3 | NP_891981.1 | |||
NM_183002.2 | 2083 | UTR 3 | NP_892114.1 | |||
XM_006720240.3 | 2083 | UTR 3 | XP_006720303.1 | |||
XM_017021606.1 | 2083 | UTR 3 | XP_016877095.1 | |||
XM_017021607.1 | 2083 | UTR 3 | XP_016877096.1 | |||
XM_017021608.1 | 2083 | UTR 3 | XP_016877097.1 | |||
XM_017021609.1 | 2083 | UTR 3 | XP_016877098.1 | |||
XM_017021610.1 | 2083 | Intron | XP_016877099.1 | |||
XM_017021611.1 | 2083 | UTR 3 | XP_016877100.1 |