Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTGGCCAGATGCACCGAACTCA[A/G]TGAGGCACTTAGAGATGAGAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601015 MIM: 609999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4709 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4709 - microRNA 4709 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPC2 - NPC intracellular cholesterol transporter 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006432.3 | 572 | UTR 3 | NP_006423.1 | |||
XM_017020930.1 | 572 | UTR 3 | XP_016876419.1 |
SYNDIG1L - synapse differentiation inducing 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |