Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTCAGAATTGTAGCTAAGTGGT[A/G]GTGGGCACATATGTGTGAAGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615094 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROX2 - prospero homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080408.2 | 4318 | Intron | NP_001073877.2 | |||
NM_001243007.1 | 4318 | Intron | NP_001229936.1 | |||
XM_005267543.3 | 4318 | Intron | XP_005267600.1 | |||
XM_017021222.1 | 4318 | UTR 3 | XP_016876711.1 |