Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGTGTTGCAAAGTGGTGACAAT[A/G]CCCCCTAGGAAGGGTTTGGATAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JDP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JDP2 - Jun dimerization protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135047.1 | 922 | Intron | NP_001128519.1 | |||
NM_001135048.1 | 922 | Intron | NP_001128520.1 | |||
NM_001135049.1 | 922 | Intron | NP_001128521.1 | |||
NM_130469.3 | 922 | Intron | NP_569736.1 | |||
XM_005267332.4 | 922 | UTR 5 | XP_005267389.1 | |||
XM_017020971.1 | 922 | Intron | XP_016876460.1 | |||
XM_017020972.1 | 922 | Intron | XP_016876461.1 | |||
XM_017020973.1 | 922 | Intron | XP_016876462.1 | |||
XM_017020974.1 | 922 | Intron | XP_016876463.1 | |||
XM_017020975.1 | 922 | Intron | XP_016876464.1 | |||
XM_017020976.1 | 922 | Intron | XP_016876465.1 |
LOC101928377 - uncharacterized LOC101928377 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |