Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTCGGGGCTAATGAGGTTTCA[A/C]CAGCCTGCTCCGTGGCTGTCTTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610865 | ||||||||||||||||||||
Literature Links: |
FLVCR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLVCR2 - feline leukemia virus subgroup C cellular receptor family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195283.1 | 1013 | Intron | NP_001182212.1 | |||
NM_017791.2 | 1013 | Missense Mutation | ACA,CCA | T,P 213 | NP_060261.2 |
LOC102724153 - uncharacterized LOC102724153 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |