Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAATAATCTTTGCTTTTTATAC[A/C]TCCCCATTCACAACCCCTTTGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612684 MIM: 605713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISM2 - isthmin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTLC2 - serine palmitoyltransferase long chain base subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004863.3 | 6308 | UTR 3 | NP_004854.1 | |||
XM_011537384.2 | 6308 | Intron | XP_011535686.1 |