Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAAACCAAAAGGATCTTCCTCA[A/G]ACTGCACTGGTCCTTCTCGGCCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610211 MIM: 603055 | ||||||||||||||||||||
Literature Links: |
SLIRP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLIRP - SRA stem-loop interacting RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267863.1 | Intron | NP_001254792.1 | ||||
NM_001267864.1 | Intron | NP_001254793.1 | ||||
NM_031210.5 | Intron | NP_112487.1 |
SNW1 - SNW domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318844.1 | Intron | NP_001305773.1 | ||||
NM_012245.2 | Intron | NP_036377.1 | ||||
XM_005267413.3 | Intron | XP_005267470.1 |