Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAGTTCTTTGAACAGGAGTAC[C/T]GTTTGTATTCTGACCAGACCAAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf178 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf178 - chromosome 14 open reading frame 178 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173978.1 | Intron | NP_001167449.1 | ||||
NM_174943.3 | Intron | NP_777603.1 |
SNW1 - SNW domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318844.1 | Intron | NP_001305773.1 | ||||
NM_012245.2 | Intron | NP_036377.1 | ||||
XM_005267413.3 | Intron | XP_005267470.1 |