Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAAAGTCAAAAAGGCTAGTAAG[C/T]AGAACAGGGCAGGTGATTCATTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCK1 - aarF domain containing kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142545.1 | Intron | NP_001136017.1 | ||||
NM_020421.3 | Intron | NP_065154.2 | ||||
XM_005267885.3 | Intron | XP_005267942.1 | ||||
XM_005267886.3 | Intron | XP_005267943.1 | ||||
XM_011536985.1 | Intron | XP_011535287.1 | ||||
XM_011536987.2 | Intron | XP_011535289.1 | ||||
XM_011536988.2 | Intron | XP_011535290.1 | ||||
XM_017021474.1 | Intron | XP_016876963.1 | ||||
XM_017021475.1 | Intron | XP_016876964.1 | ||||
XM_017021476.1 | Intron | XP_016876965.1 | ||||
XM_017021477.1 | Intron | XP_016876966.1 |