Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAACATCTTCCTCCCTGATAGC[C/G]CATGTCTTTCTATGTTTCCTTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIO2 - deiodinase, iodothyronine, type II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000793.5 | 3545 | UTR 3 | NP_000784.2 | |||
NM_001007023.4 | 3545 | UTR 3 | NP_001007024.1 | |||
NM_001242502.2 | 3545 | UTR 3 | NP_001229431.1 | |||
NM_001242503.2 | 3545 | UTR 3 | NP_001229432.1 | |||
NM_001324462.1 | 3545 | UTR 3 | NP_001311391.1 | |||
NM_013989.4 | 3545 | UTR 3 | NP_054644.1 |