Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAATGAGTTATGGCCCTCTGC[G/T]CTTTCAAATCTCATCTTAAATCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC7B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC7B - tetratricopeptide repeat domain 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010854.1 | Intron | NP_001010854.1 | ||||
NM_001320421.1 | Intron | NP_001307350.1 | ||||
XM_005267367.1 | Intron | XP_005267424.1 | ||||
XM_011536497.1 | Intron | XP_011534799.1 | ||||
XM_017021044.1 | Intron | XP_016876533.1 | ||||
XM_017021045.1 | Intron | XP_016876534.1 | ||||
XM_017021046.1 | Intron | XP_016876535.1 | ||||
XM_017021047.1 | Intron | XP_016876536.1 |