Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCATAGTTTTCGAGTGAGTGTGA[A/G]TGATTCATAGTTTTTTATATATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD7 - BTB domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002860.3 | 4252 | UTR 3 | NP_001002860.2 | |||
NM_001289133.1 | 4252 | UTR 3 | NP_001276062.1 | |||
NM_018167.4 | 4252 | Intron | NP_060637.1 | |||
XM_011536939.2 | 4252 | Intron | XP_011535241.1 | |||
XM_017021438.1 | 4252 | Intron | XP_016876927.1 |