Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGCGGTCCGGGTGGTCGGTGA[C/T]TGCCTCCCCGAGTGTCGGGCACTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCL1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCL1A - T-cell leukemia/lymphoma 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098725.1 | 161 | Missense Mutation | ATC,GTC | I,V 11 | NP_001092195.1 | |
NM_021966.2 | 161 | Missense Mutation | ATC,GTC | I,V 11 | NP_068801.1 | |
XM_017021676.1 | 161 | Missense Mutation | ATC,GTC | I,V 11 | XP_016877165.1 | |
XM_017021677.1 | 161 | Missense Mutation | ATC,GTC | I,V 11 | XP_016877166.1 |