Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGAGGGCAGAGGTCCCATAGGCC[C/T]CTTCCCTCCCAGAGTTTCCCCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf2 - chromosome 14 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD9 - tudor domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153046.2 | Intron | NP_694591.2 | ||||
XM_005267309.3 | Intron | XP_005267366.1 | ||||
XM_006720019.3 | Intron | XP_006720082.1 | ||||
XM_006720020.3 | Intron | XP_006720083.1 | ||||
XM_011536397.2 | Intron | XP_011534699.1 | ||||
XM_011536398.2 | Intron | XP_011534700.1 | ||||
XM_011536400.2 | Intron | XP_011534702.1 | ||||
XM_011536401.2 | Intron | XP_011534703.1 | ||||
XM_011536402.2 | Intron | XP_011534704.1 | ||||
XM_017020955.1 | Intron | XP_016876444.1 |