Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCAAGTGCCAGATCCTCAGCTC[C/T]CACGGCTGCGCCGGACACCTGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRF1 - BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242786.1 | 3197 | UTR 3 | NP_001229715.1 | |||
NM_001242787.1 | 3197 | UTR 3 | NP_001229716.1 | |||
NM_001242788.1 | 3197 | UTR 3 | NP_001229717.1 | |||
NM_001242789.1 | 3197 | UTR 3 | NP_001229718.1 | |||
NM_001242790.1 | 3197 | Intron | NP_001229719.1 | |||
NM_001519.3 | 3197 | UTR 3 | NP_001510.2 | |||
NM_145685.2 | 3197 | UTR 3 | NP_663718.1 | |||
XM_005267561.3 | 3197 | UTR 3 | XP_005267618.1 | |||
XM_005267563.3 | 3197 | UTR 3 | XP_005267620.1 | |||
XM_006720123.2 | 3197 | UTR 3 | XP_006720186.1 | |||
XM_011536672.2 | 3197 | Intron | XP_011534974.1 | |||
XM_011536673.1 | 3197 | UTR 3 | XP_011534975.1 | |||
XM_011536674.1 | 3197 | UTR 3 | XP_011534976.1 |