Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600162 MIM: 182279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PWAR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PWAR5 - Prader Willi/Angelman region RNA 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PWARSN - Prader Willi/Angelman region RNA, SNRPN neighbor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD107 - small nucleolar RNA, C/D box 107 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD108 - small nucleolar RNA, C/D box 108 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD64 - small nucleolar RNA, C/D box 64 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRPN - small nuclear ribonucleoprotein polypeptide N | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNURF - SNRPN upstream reading frame | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |